MilliporeSigma
  • SZ-685C, a marine anthraquinone, is a potent inducer of apoptosis with anticancer activity by suppression of the Akt/FOXO pathway.
SZ-685C, a marine anthraquinone, is a potent inducer of apoptosis with anticancer activity by suppression of the Akt/FOXO pathway.
British journal of pharmacology (2010-02-05)
Gui'e Xie, Xun Zhu, Qing Li, Minghui Gu, Zhenjian He, Jueheng Wu, Jun Li, Yongcheng Lin, Mengfeng Li, Zhigang She, Jie Yuan
ABSTRACT
The aims of this study were to investigate the anti-cancer activity of SZ-685C, an anthracycline analogue isolated from marine-derived mangrove endophytic fungi, and to explore the molecular mechanisms underlying such activity. The effect of SZ-685C on the viability of cancer cell lines was investigated using the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide (MTT) assay. SZ-685C-induced apoptosis was assessed by Annexin V-fluorescein isothiocyanate/propidium iodide staining, terminal deoxynucleotidyl transferase-mediated dUTP nick end labelling assay and analysis of caspase activation. The effect of SZ-685C on the Akt/FOXO pathway was studied using Western blotting analysis, and the in vivo anti-tumour efficacy was examined in an MDA-MB-435 breast cancer xenograft model. SZ-685C suppressed the proliferation of six cancer cell lines derived from human breast cancer, prostate cancer, glioma and hepatoma (IC(50) values ranged from 3.0 to 9.6 microM) and the growth of breast cancer xenografts in mice. SZ-685C had a direct apoptosis-inducing effect through both the extrinsic and intrinsic apoptotic pathways, as shown by activation of caspase-8 and 9 as well as effector caspase-3 and poly (ADP-ribose) polymerase. Phosphorylation of Akt and its downstream effectors, forkhead box protein O1 and forkhead box protein O3a, was down-regulated in SZ-685C-treated cancer cells. Furthermore, the pro-apoptotic protein Bim was up-regulated by SZ-685C treatment consistent with FOXO dephosphorylation. SZ-685C could induce apoptosis through the Akt/FOXO pathway, which consequently leads to the observed anti-tumour effect both in vitro and in vivo. Our data suggest that SZ-685C may be a potentially promising Akt inhibitor and anti-cancer drug candidate.
Product Description
Supelco
Melting point standard 283-286°C, analytical standard
Sigma-Aldrich
Anthraquinone, 97%