MilliporeSigma
  • Characterization of microbial communities in the aqueous phase of a constructed model wetland treating 1,2-dichloroethene-contaminated groundwater.
Characterization of microbial communities in the aqueous phase of a constructed model wetland treating 1,2-dichloroethene-contaminated groundwater.
FEMS microbiology ecology (2010-01-27)
Gwenaël Imfeld, Cristian Estop Aragonés, Ingo Fetzer, Eva Mészáros, Simone Zeiger, Ivonne Nijenhuis, Marcell Nikolausz, Sylvain Delerce, Hans H Richnow
ABSTRACT
The dynamics and composition of microbial communities in the aqueous phase of a model wetland supplied with cis- and trans-1,2-dichloroethenes (DCE)-contaminated groundwater was characterized. PCR-denaturing gradient gel electrophoresis analysis of water samples obtained from different parts of the wetland revealed that changes of the bacterial community structure coincided with a succession of the hydrochemical conditions in the wetland, from oxic towards anoxic conditions. During this transition phase, the appearance of vinyl chloride and ethene correlated with the presence of putative dechlorinating bacteria (Dehalococcoides spp., Geobacter spp. and Dehalobacter spp.). Additionally, a shift of the DCE isotopic composition indicated the progressive prevalence of reductive dechlorination in the wetland. Although the DCE degradation processes varied over time, biodegradation activity was maintained in the wetland system. 16S rRNA gene libraries revealed that Proteobacteria accounted for >50% of 16S rRNA genes clone libraries, whereas approximately 17% of the sequences from the wetland were related to sulphate reducers. Based on a multiple-method approach, this study illustrates the linkage between microbial community dynamics and composition, changes of hydrochemical conditions and processes of DCE degradation in a wetland system.
Product Description
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Supelco
cis-1,2-Dichloroethene, analytical standard