MilliporeSigma
  • A possibility of nutriceuticals as an anti-aging intervention: activation of sirtuins by promoting mammalian NAD biosynthesis.
A possibility of nutriceuticals as an anti-aging intervention: activation of sirtuins by promoting mammalian NAD biosynthesis.
Pharmacological research (2010-01-21)
Shin-Ichiro Imai
ABSTRACT
Aging science has recently drawn much attention, and discussions on the possibility of anti-aging medicine have multiplied. One potential target for the development of anti-aging drugs is the SIR2 (silent information regulator 2) family of NAD-dependent deacetylases/ADP-ribosyltransferases, called "sirtuins." Sirtuins regulate many fundamental biological processes in response to a variety of environmental and nutritional stimuli. In mammals, the mammalian SIR2 ortholog SIRT1 has been most studied, and small molecule SIRT1 activators (STACs), including a plant-derived polyphenolic compound resveratrol, have been developed. On the other hand, sirtuin activity is regulated by NAD biosynthetic pathways, and nicotinamide phosphoribosyltransferase (NAMPT) plays a critical role in the regulation of mammalian sirtuin activity. Recent studies have provided a proof of concept for the idea that nicotinamide mononucleotide (NMN), the NAMPT reaction product, can be used as a nutriceutical to activate SIRT1 activity. Based on these recent findings, the possibility of sirtuin-targeted nutriceutical development will be discussed.
Sigma-Aldrich
β-Nicotinamide mononucleotide, ≥95% (HPLC)