MilliporeSigma
  • Analysis of supercoiled DNA by agarose gel electrophoresis using low-conducting sodium threonine medium.
Analysis of supercoiled DNA by agarose gel electrophoresis using low-conducting sodium threonine medium.
Analytical biochemistry (2010-01-21)
Tomomi Ishido, Mitsuru Ishikawa, Ken Hirano
ABSTRACT
We describe a new low-ionic-strength sodium threonine (STh) medium with the advantage of avoiding relative DNA band migration changes following electrophoresis of supercoiled DNA in agarose gel when substituted for the standard conductive medium of TBE (Tris-boric acid-ethylenediaminetetraacetic acid [EDTA]) or TAE (Tris-acetic acid-EDTA) or the low-ionic-strength sodium boric acid medium. Low-ionic-strength STh medium provided better resolution, less heat generation, and prevention of relative migration order changes among linear, covalently closed circular-, and open circular-formed DNA in the range of 2-10 kilobase pairs in 1% agarose gel electrophoresis.
Sigma-Aldrich
Borax Anhydrous, puriss. p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Borax Anhydrous, BioUltra, anhydrous, ≥99%
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Sodium tetraborate, 99%