MilliporeSigma
  • Relevance of 1,2,5,6,9,10-hexabromocyclododecane diastereomer structure on partitioning properties, column-retention and clean-up procedures.
Relevance of 1,2,5,6,9,10-hexabromocyclododecane diastereomer structure on partitioning properties, column-retention and clean-up procedures.
Journal of chromatography. A (2010-01-19)
Espen Mariussen, Marianne Haukås, Hans Peter H Arp, Kai-Uwe Goss, Anders Borgen, Torkjel M Sandanger
ABSTRACT
To optimize clean-up procedures for the analysis of alpha-, beta-, and gamma-hexabromocyclododecanes (HBCD) in environmental and biological extracts, their retention behavior on silica gel and florisil was investigated using diverse mobile-phase solvents and accounting for matrix effects. The beta-diastereomer, relative to the alpha- and gamma-diastereomers, is substantially retained on both florisil and silica gel regardless of the solvent used. The beta-diastereomer is therefore prone to undergo selective loss during clean-up. This sequence is counterintuitive to sequences based on reverse-phase chromatography with a C18-column, in which the alpha- (and not the beta-) isomer is eluted first when using a polar solvent. There has been some discrepancy regarding the structures of these diastereomers in the literature, with structures based on X-ray crystallography only becoming recently available. Based on these X-ray crystal structures, physical-chemical properties (the octanol-water partitioning constant, the Henry's law constant, subcooled liquid vapour pressures and subcooled liquid water solubilities) of the HBCD diastereomers were estimated using the quantum-chemistry based software COSMOtherm, and were found to differ from previously calculated values using different structures (e.g. log K(aw) for alpha-, beta-, and gamma-HBCD are here estimated to be -8.3, -9.3 and -8.2 respectively). Hypothesis relating differences in structure to physical-chemical properties and retention sequences are presented. The extra retention of the beta-diastereomer on silica gel and florisil is likely because it can form both greater specific (i.e. polar) and non-specific (i.e. non-polar) interactions with surfaces than the other diastereomers. Non-specific interactions can also account for the counter-intuitive elution orders with C(18)-reverse-phase chromatography. These results indicate that care should be taken when isolating HBCDs and other molecular diastereomers from environmental and biological samples, and that reported concentrations of beta-HBCD in the literature may be negatively biased.
Product Description
Supelco
Florisil® Adsorbent for Chromatography, 30-60 mesh
Supelco
Florisil® Adsorbent for Chromatography, 60-100 mesh
Supelco
Florisil®, <200 mesh, fine powder
Supelco
Florisil®, for the determination of hydrocarbon acc. to ISO 9377-2
Supelco
Florisil® PR
Supelco
Florisil®, 60-100 mesh, coarse powder
Supelco
Florisil®, 100-200 mesh, fine powder
Sigma-Aldrich
Florisil®, 100-200 mesh