MilliporeSigma
  • Synthesis of oxepines and 2-branched pyranosides from a d-glucal-derived gem-dibromo-1,2-cyclopropanated sugar.
Synthesis of oxepines and 2-branched pyranosides from a d-glucal-derived gem-dibromo-1,2-cyclopropanated sugar.
The Journal of organic chemistry (2010-01-14)
Russell J Hewitt, Joanne E Harvey
ABSTRACT
The conversion of cyclopropane-fused carbohydrates into oxepines is an attractive method for accessing diverse members of the septanoside family of carbohydrate mimetics. 2-Bromooxepines are obtained through silver(I)-promoted thermal ring expansion of a d-glucal-derived gem-dihalocyclopropanated sugar. In contrast, cyclopropane ring cleavage under basic conditions leads to 2-C-branched pyranosides, not the 2-bromooxepine structures assigned in an earlier report.
Sigma-Aldrich
D-Glucal, 96%