MilliporeSigma
  • Determination of para red, Sudan dyes, canthaxanthin, and astaxanthin in animal feeds using UPLC.
Determination of para red, Sudan dyes, canthaxanthin, and astaxanthin in animal feeds using UPLC.
Journal of chromatographic science (2010-01-09)
Xiaolin Hou, Yonggang Li, Guojuan Wu, Lei Wang, Miao Hong, Yongnin Wu
ABSTRACT
A simple high-performance liquid chromatography method was developed for quantitative determination of para red, Sudan I, Sudan II, Sudan III, Sudan IV, canthaxanthin, and astaxanthin in feedstuff. The sample was extracted using acetonitrile and cleaned up on a C(18) SPE column. The residues were analyzed using ultra-performance liquid chromatography coupled to a diode array detector at 500 nm. The mobile phase was acetonitrile-formic acid-water with a gradient elution condition. The external standard curves were calibrated. The mean recoveries of the seven colorants were 62.7-91.0% with relative standard deviation 2.6-10.4% (intra-day) and 4.0-13.2% (inter-day). The detection limits were in the range of 0.006-0.02 mg/kg.
Product Description
Supelco
Sudan III, analytical standard
Sigma-Aldrich
Sudan III, BioXtra, certified by the Biological Stain Commission
Sigma-Aldrich
Sudan III, Technical grade
Supelco
Sudan IV, analytical standard
Sigma-Aldrich
Sudan IV, Certified Biological Stain Commission,Dye content ≥80 %
Sigma-Aldrich
Sudan II, Dye content 90 %
Sigma-Aldrich
Sudan I, Dye content ≥95 %
Supelco
Sudan I, analytical standard
Sigma-Aldrich
Sudan IV, BioXtra, certified by the Biological Stain Commission
Supelco
Sudan II, analytical standard