MilliporeSigma
  • Syntheses of carnosic acid and carnosol, anti-oxidants in Rosemary, from pisiferic acid, the major constituent of Sawara.
Syntheses of carnosic acid and carnosol, anti-oxidants in Rosemary, from pisiferic acid, the major constituent of Sawara.
Chemical & pharmaceutical bulletin (2010-01-05)
Masahiro Tada, Tomoyuki Ohkanda, Jun Kurabe
ABSTRACT
Carnosic acid (2), a major anti-oxidant in rosemary (Rosmarinus officinalis), was synthesized from pisiferic acid (1), the major constituent of Sawara (Chamaecyparis pisifera), via ortho-oxidation of the phenol using meta-chlorobenzoyl peroxide (mCBPO), chloroacetyl meta-chlorobenzoyl peroxide (CAMCBPO) or 2-iodoxybenzoic acid (IBX). Carnosol (3), another anti-oxidant in rosemary, was synthesized from carnosic acid by oxidation with silver oxide. Potent antibacterial activities against Propionibacterium acnes (ATCC 6919) (minimum inhibitory concentration (MIC) mug/ml) and Staphylococcus aureus ME/GM/TC Resistant (ATCC 33592) (MIC mug/ml) of carnosic acid and carnosol were reported.
Sigma-Aldrich
Carnosol
Supelco
Carnosol, analytical standard