MilliporeSigma
  • Photodissociation of cis-, trans-, and 1,1-dichloroethylene in the ultraviolet range: characterization of Cl((2)P(J)) elimination.
Photodissociation of cis-, trans-, and 1,1-dichloroethylene in the ultraviolet range: characterization of Cl((2)P(J)) elimination.
The journal of physical chemistry. A (2009-12-22)
Linqiang Hua, Xiaopeng Zhang, Wei-Bin Lee, Meng-Hsuan Chao, Bing Zhang, King-Chuen Lin
ABSTRACT
By using photofragment velocity imaging detection coupled with a (2 + 1) resonance-enhanced multiphoton ionization technique, the elimination channel of spin-orbit chlorine atoms in photodissociation of cis-, trans-, and 1,1-dichloroethylene at two photolysis wavelengths of 214.5 and 235 nm is investigated. Translational energy and angular distributions of Cl((2)P(J)) fragmentation are acquired. The Cl((2)P(J)) fragments are produced by two competing channels. The fast dissociation component with higher translational energy is characterized by a Gaussian distribution, resulting from a curve crossing of the initially excited (pi, pi*) state to nearby repulsive (pi, sigma*) and/or (n, sigma*). In contrast, the slow component with a lower translational energy is characterized by a Boltzmann distribution, which dissociates on the vibrationally hot ground state relaxed from the (pi, pi*) state via internal conversion. cis-C(2)H(2)Cl(2) is found to have a larger branching of Boltzmann component than the other two isomers. The fraction of available energy partitioning into translation increases along the trend of cis- < trans- < 1,1-C(2)H(2)Cl(2). This trend may be fitted by a rigid radical model and interpreted by means of a torque generated during the C-Cl bond cleavage. The anisotropy parameters are determined, and the transition dipole moments are expected to be essentially along the C horizontal lineC bond axis. The results are also predicted theoretically. The relative quantum yields of Cl((2)P(J)) have a similar value for the three isomers at the two photolysis wavelengths.
Product Description
Supelco
1,1-Dichloroethene solution, certified reference material, 1000 μg/mL in methanol
Supelco
1,1-Dichloroethylene, analytical standard