MilliporeSigma
  • 2,2,2-trichloroethanol activates a nonclassical potassium channel in cerebrovascular smooth muscle and dilates the middle cerebral artery.
2,2,2-trichloroethanol activates a nonclassical potassium channel in cerebrovascular smooth muscle and dilates the middle cerebral artery.
The Journal of pharmacology and experimental therapeutics (2009-12-04)
Nikhil K Parelkar, Neerupma Silswal, Kirsten Jansen, Joshua Vaughn, Robert M Bryan, Jon Andresen
ABSTRACT
Trichloroacetaldehyde monohydrate [chloral hydrate (CH)] is a sedative/hypnotic that increases cerebral blood flow (CBF), and its active metabolite 2,2,2-trichloroethanol (TCE) is an agonist for the nonclassical two-pore domain K(+) (K(2P)) channels TREK-1 and TRAAK. We sought to determine whether TCE dilates cerebral arteries in vitro by activating nonclassical K(+) channels. TCE dilated pressurized and perfused rat middle cerebral arteries (MCAs) in a manner consistent with activation of nonclassical K(+) channels. Dilation to TCE was inhibited by elevated external K(+) but not by an inhibitory cocktail (IC) of classical K(+) channel blockers. Patch-clamp electrophysiology revealed that, in the presence of the IC, TCE increased whole-cell currents and hyperpolarized the membrane potential of isolated MCA smooth muscle cells. Heating increased TCE-sensitive currents, indicating that the activated channel was thermosensitive. Immunofluorescence in sections of the rat MCA demonstrated that, like TREK-1, TRAAK is expressed in the smooth muscle of cerebral arteries. Isoflurane did not, however, dilate the MCA, suggesting that TREK-1 was not functional. These data indicate that TCE activated a nonclassical K(+) channel with the characteristics of TRAAK in rat MCA smooth-muscle cells. Stimulation of K(+) channels such as TRAAK in cerebral arteries may therefore explain in part how CH/TCE increases CBF.
Sigma-Aldrich
2-Chloroethanol, 99%
Sigma-Aldrich
2,2,2-Trichloroethanol, ReagentPlus®, ≥99%