MilliporeSigma
  • Synthesis and AChE inhibitory activity of new chiral tetrahydroacridine analogues from terpenic cyclanones.
Synthesis and AChE inhibitory activity of new chiral tetrahydroacridine analogues from terpenic cyclanones.
European journal of medicinal chemistry (2009-12-04)
Diego dos Santos Pisoni, Jessé Sobieski da Costa, Douglas Gamba, Cesar Liberato Petzhold, Antonio César de Amorim Borges, Marco Antonio Ceschi, Paula Lunardi, Carlos Alberto Saraiva Gonçalves
ABSTRACT
This work describes the enantioselective synthesis of a new series of terpenic chiral 9-aminotetrahydroacridine analogues. Several chiral ketones were synthesized from natural monoterpenes in an optically active form and subjected to the cyclodehydration reactions with anthranilonitrile in the presence of BF(3).Et(2)O as catalyst. The 9-aminotetrahydroacridine analogues were tested as acetylcholinesterase (AChE) inhibitors. Based on qualitative structure-activity relationship some trends are suggested.
Product Description
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), for GC derivatization, LiChropur