MilliporeSigma
[Chemical constituents of Lobelia chinensis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2009-12-01)
Jinglan Han, Fengling Zhang, Zhihong Li, Guanhua Du, Hailin Qin
ABSTRACT
To study the chemical constituents from Lobelia chinensis. The constituents were extracted with 95% EtOH, partitioned with different solvents, and isolated and purified by silica gel column chromatography and crystallization, their structures were elucidated by physico-chemical properties and spectroscopic data. Seven compounds were isolated from the titled plant. They were identified as 5-hydroxy-4'-methoxyflavone-7-O-rutinoside (1), n-butyl-O-beta-D-fructopyranoside (2), 5,7-dimethoxycoumarin (3), cirsiumaldehyde (4), diosmetin (5), 5-hydroxymethyl-2-furancarboxaldehyde (6), and 6-hydroxy-7-methoxycoumarin (7). Compounds 2-4, 6, and 7 were obtained from the titled plant for the first time.
Sigma-Aldrich
5,7-Dimethoxycoumarin, 98%