MilliporeSigma
  • Penetration pathways induced by low-frequency sonophoresis with physical and chemical enhancers: iron oxide nanoparticles versus lanthanum nitrates.
Penetration pathways induced by low-frequency sonophoresis with physical and chemical enhancers: iron oxide nanoparticles versus lanthanum nitrates.
The Journal of investigative dermatology (2009-11-27)
Sang Eun Lee, Ki Ju Choi, Gopinathan K Menon, Hyun Jung Kim, Eung Ho Choi, Sung Ku Ahn, Seung Hun Lee
ABSTRACT
Low-frequency sonophoresis (LFS) has been shown to disrupt the structure of stratum corneum (SC) lipid bilayers and enhance SC permeability. In this study, we examined the penetration pathway of lanthanum nitrate (LaNO(3)) tracer in viable epidermis after combined treatment of LFS and tape stripping (TS), as a physical enhancer, or oleic acid (OA) application, as a chemical enhancer, using transmission electron microscopy (TEM). As a positive control, we visualized the passive diffusion pathway of LaNO(3) and iron oxide (Fe(3)O(4)) nanoparticles after the incision of hairless mouse skin. Next, we applied LFS immediately after TS or OA application and visualized the penetration pathway of LaNO(3). Each treatment showed restricted penetration to the SC-stratum granulosum (SG) interface or upper SG layer. However, the additional application of LFS induced diffuse intracellular distribution of LaNO(3) throughout the viable epidermis. Quantitative analysis also revealed that combined treatment significantly increases LaNO(3) penetration into viable epidermis when compared with each treatment. Our ultrastructural findings show the synergistic effect of LFS and TS or OA application on transdermal drug delivery. We also found that this combined treatment enhances the penetration of LaNO(3) through the viable epidermis through an intracellular pathway.
Sigma-Aldrich
Lanthanum(III) nitrate hexahydrate, 99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) nitrate hexahydrate, 99.999% trace metals basis