MilliporeSigma
  • Experimental and clinical studies on Rifacinna--the new effective antituberculous drug (review).
Experimental and clinical studies on Rifacinna--the new effective antituberculous drug (review).
Recent patents on anti-infective drug discovery (2009-11-26)
Dimova Velichka, Atanasova Ivana, Tomioka Haruaki, Sato Katsumasa, Reddy Venkata, Geeta Nadadhur, Daneluzzi Donna, Kantardjiev Todor, Dhople Arvind, Feschenko Yurii, Yashina Ljudmila, Andrei Toumanov, Zhivkova Zvetana, Sano Chiaki
ABSTRACT
A new rifamycin derivative 3-(4-cinnamyl-piperazinyl iminomethyl) rifamycin SV (T9) and its sodium salt (T11, Rifacinna((R))) were in vitro, in vivo, toxicologically and clinically investigated in comparison with rifampicin, rifapentine, rifabutin, rifalazil. Our experiments showed that Rifacinna exhibits excellent in vitro activity against Gram(+), Gram (-) aerobic, anaerobic pathogens and mycobacteria. Rifacinna is active against Staphylococcus, Streptococcus spp. including MRSA, with MIC90- 0.06-0.5 mg/L; against Gram(+), Gram (-) anaerobes with MIC90 0.5 - 1 mg/L; against Mycobacterium tuberculosis (MTB) with MIC90 0.062 mg/L; against MDR resistant MTB (25%-30 %) and Mycobacterium avium complex (MAC) strains with MICs 0.6-1.0 mg/L. It shows high intraphagocytic activity against MAC strains (0.06 0.125mg/L). Single daily dose 10 mg/kg provides complete erradication of mycobacteria in experimental generalized tuberculosis. Pharmacological studies established: excellent pharmacokinetic profile following single oral dose 10mg/kg Tmax 5-6 h, C(max) 5-9 mg/L, T1/2 33-34 h; low toxicity; no teratogenic and embryotoxic effects. The clinical study of Rifacinna shows higher therapeutic efficacy than Rifampicin in patients with infiltrative, disseminated and cavitary form of pulmonary tuberculosis, good tolerability and safety profile. Some of the recent patents related to the treatment of tuberculosis are discussed in this review article.
Sigma-Aldrich
Rifamycin SV sodium salt, potency: ≥ 900 IU/mg (anhydrous basis)
Rifamycin sodium, European Pharmacopoeia (EP) Reference Standard