MilliporeSigma
  • Antibacterial and antifungal activity of a soda-lime glass containing copper nanoparticles.
Antibacterial and antifungal activity of a soda-lime glass containing copper nanoparticles.
Nanotechnology (2009-11-13)
L Esteban-Tejeda, F Malpartida, A Esteban-Cubillo, C Pecharromán, J S Moya
ABSTRACT
A low melting point soda-lime glass powder containing copper nanoparticles with high antibacterial (against gram-positive and gram-negative bacteria) and antifungal activity has been obtained. Sepiolite fibres containing monodispersed copper nanoparticles (d(50) approximately 30 +/- 5 nm) were used as the source of the copper nanoparticles. The observed high activity of the obtained glass powder, particularly against yeast, has been explained by considering the inhibitory synergistic effect of the Ca(2+) lixiviated from the glass on the growth of the colonies.
Sigma-Aldrich
Soda lime, p.a., with indicator, pellets
Sigma-Aldrich
Soda lime, ACS reagent, granular, +100 mesh particle size