MilliporeSigma
  • Tetranuclear vanadium complex, (VO)(4)(hpic)(4): a recyclable catalyst for oxidation of benzyl alcohols with molecular oxygen.
Tetranuclear vanadium complex, (VO)(4)(hpic)(4): a recyclable catalyst for oxidation of benzyl alcohols with molecular oxygen.
Dalton transactions (Cambridge, England : 2003) (2009-11-04)
Shintaro Kodama, Yukihiro Ueta, Jun Yoshida, Akihiro Nomoto, Shigenobu Yano, Michio Ueshima, Akiya Ogawa
ABSTRACT
Vanadium(iv) complexes bearing 3-hydroxypicolinic acid (H(2)hpic) as ligands, VO(Hhpic)(2) (1) and the cyclic tetramer (VO)(4)(hpic)(4) (2), have excellent catalytic ability for the oxidation of a variety of primary and secondary benzyl alcohols with molecular oxygen in acetonitrile or protic solvents such as ethanol and water, affording the corresponding aldehydes and ketones, respectively. Construction of multi-nuclear complexes by the selection of ligands attains higher turnover numbers and recycling of the catalyst.
Product Description
Supelco
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%