MilliporeSigma
  • Direct-oxidative DNA damage and apoptosis induction in different human respiratory cells exposed to low concentrations of sodium chromate.
Direct-oxidative DNA damage and apoptosis induction in different human respiratory cells exposed to low concentrations of sodium chromate.
Journal of applied toxicology : JAT (2009-10-20)
Delia Cavallo, Cinzia Lucia Ursini, Anna Maria Fresegna, Aureliano Ciervo, Raffaele Maiello, Bruna Rondinone, Velia D'Agata, Sergio Iavicoli
ABSTRACT
The mechanism of Cr(VI) genotoxicity has still not been elucidated. We used Fpg-modified comet assay to assess direct-oxidative DNA damage on human lung (A549) and bronchial (BEAS-2B) cells exposed to 0.1, 0.5, 1.0 and 10 microm sodium chromate for 0.5, 1 and 4 h. Moreover we evaluated apoptosis by morphological analysis and caspase-3 activity, also after 24 h. On A549 cells a time-dependent DNA damage, expressed as tail DNA%, beginning from 0.5 microm was found. For oxidative DNA damage an induction after 30 min to 0.5 microm decreasing with time, and a time-dependent increase at 10 microm was found, indicating for low Cr(VI) concentration the oxidative stress as the first event followed by direct DNA damage and for the highest concentration a time-dependent increase in oxidative DNA damage. On BEAS-2B cells DNA damage was induced within 1 h at 0.5-10 microm, without changes with time, showing that BEAS-2B cells are able to resist to Cr(VI) genotoxicity. Early oxidative DNA damage at 0.1 microm decreasing with time was also found. Significant apoptosis was observed by morphological analysis in A549 cells and to a lower extent in BEAS-2B at 10 microm. The exposure to 10 microm induced caspase-3 activity after 4 h in BEAS-2B and after 24 h in A549 cells. The findings show a higher responsiveness of A549 cells to genotoxic effect of Cr(VI) and early transient oxidative DNA damage in BEAS-2B. The results emphasize the suitability of this experimental model to evaluate the early genotoxic response of different cells to non-cytotoxic concentrations of Cr(VI) on target organ.
Sigma-Aldrich
Sodium chromate, 98%
Sigma-Aldrich
Sodium chromate tetrahydrate, 99%