MilliporeSigma
  • The activity and compatibility of the antibiotic tiamulin with other drugs in poultry medicine--A review.
The activity and compatibility of the antibiotic tiamulin with other drugs in poultry medicine--A review.
Poultry science (2009-10-17)
K M S Islam, U Klein, D G S Burch
ABSTRACT
Tiamulin hydrogen fumarate is a semisynthetic derivative of the diterpene antibiotic pleuromutilin used in poultry medicine to treat mainly Mycoplasma- and Brachyspira-related diseases. Its use over 30 yr has not generally increased the development of resistance to these pathogens but occasionally resistant isolates are encountered. Tiamulin administered at therapeutic levels is relatively quickly absorbed, metabolized in the liver, and eliminated from the body of the bird after a withdrawal period of 72 h, and as a result, meat products can be safely consumed. A zero withdrawal period for eggs has been granted in several European Union states. When administered with different drugs, tiamulin has been shown to have an enhanced activity with the tetracyclines. There is a strong interaction, even death, with the ionophore anticoccidials monensin, narasin, and salinomycin when tiamulin is used at therapeutic levels, but this is dose-related and low doses do not interact. It is thought to be caused by the preferential metabolism of tiamulin in the liver resulting in a build up of the ionophore leading to clinical signs of overdosage. Tiamulin shows a milder interaction, such as temporary growth depression, with maduramicin and semduramicin but is compatible with lasalocid. Although tiamulin shows small benefits in improving performance in healthy animals, its main production benefit is in the face of infection, as a true therapeutic antibiotic.
Product Description
Supelco
Tiamulin, VETRANAL®, analytical standard
Supelco
Tiamulin fumarate, VETRANAL®, analytical standard
Tiamulin hydrogen fumarate, European Pharmacopoeia (EP) Reference Standard