MilliporeSigma
  • Supramolecular chirality and chiral inversion of tetraphenylsulfonato porphyrin assemblies on optically active polylysine.
Supramolecular chirality and chiral inversion of tetraphenylsulfonato porphyrin assemblies on optically active polylysine.
The journal of physical chemistry. B (2009-10-16)
Li Zhang, Minghua Liu
ABSTRACT
The self-assembly and induced supramolecular chirality of a dianionic meso-tetraphenylsulfonato porphyrin (TPPS) on the optically active polylysine has been investigated. Our research has confirmed that in the presence of poly(l-lysine) (PLL) or poly(d-lysine) (PDL), TPPS could form both H and J aggregates and the exciton type Cotton effect was induced in the corresponding absorption bands of H and J-aggregates. We have further revealed that the induced chirality of the H-band always followed the chirality of PLL or PDL, while the sign of the exciton couplet in the J-band could be the same as or opposite to that of the H-band depending on the mixing sequence and the ratio of PLL or PDL to TPPS (P/T ratio). At a P/T ratio less than 4, both the J and H aggregates showed the same symbolic CD signal. At a P/T ratio larger than 4, the opposite sign of the exciton couplet was observed for H and J-bands when TPPS was added into PLL, while the same sign was obtained when PLL was added into TPPS. Interestingly, the J-band with the same sign as that of the H-band can be inverted into the opposite sign under heating. A mechanism relating to the dynamic and thermodynamic formation of the chiral aggregates in the presence of PLL or PDL was proposed.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%