Merck
  • Aqueous-core lipid nanocapsules for encapsulating fragile hydrophilic and/or lipophilic molecules.
Aqueous-core lipid nanocapsules for encapsulating fragile hydrophilic and/or lipophilic molecules.
Langmuir : the ACS journal of surfaces and colloids (2009-10-01)
Nicolas Anton, Patrick Saulnier, Cédric Gaillard, Emilien Porcher, Sandy Vrignaud, Jean-Pierre Benoit
ABSTRACT
This paper presents the original method of producing aqueous-core lipid nanocapsule, able to encapsulate both hydrophilic and lipophilic species with relevant yields. The scope offered by the integration of both hydrophilic and lipophilic molecules within the same colloidal objects make this method a prime candidate for the numerous anticancer therapies for which chemotherapy frequently requires the association of several molecules. The proof of concept study is proposed here (i) describing in detail the formulation of such objects and their characterization (TEM, cryo-TEM, soft particle analysis), (ii) showing the encapsulation of hydrophilic species alone, using examples of very different molecular weights, such as a dye (methylene blue) and a protein (BSA-isothycianate fluorecein labeled), and (iii) showing the simultaneous encapsulation of hydrophilic (methylene blue) and lipophilic (also a dye, red Sudan III) species.
Sigma-Aldrich
Sudan III, certified by the Biological Stain Commission, BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sudan III, Technical grade
SKU
Pack Size
Availability
Price
Quantity
Supelco
Sudan III, analytical standard