MilliporeSigma
  • Revision of analytical conditions for determining ligand molecules specific to soft metal ions using dequenching of copper(I)-bathocuproine disulfonate as a detection system.
Revision of analytical conditions for determining ligand molecules specific to soft metal ions using dequenching of copper(I)-bathocuproine disulfonate as a detection system.
Analytical chemistry (2009-09-30)
Shinya Ogawa, Rina Ichiki, Mitsuru Abo, Etsuro Yoshimura
ABSTRACT
Reoptimization of analytical conditions was performed for a high-performance liquid chromatographic (HPLC) detection system for Cu(I) chelators based on the dequenching of Cu(I)-bathocuproine disulfonate complexes that occurs in the presence of Cu(I) chelators. The revision corrects for emission and excitation wavelengths that were in fact second-order light of the actual optimal wavelengths and for the composition of the postcolumn solution. These revisions resulted in an order of magnitude decrease in detection limits of phytochelatins, a class of cysteine-rich, heavy metal-binding peptides. The revised technique is capable of phytochelatin quantitation at femtomole quantities.
Sigma-Aldrich
Bathocuproinedisulfonic acid disodium salt, for spectrophotometric det. of Cu, Fe