MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Open-sandwich enzyme immunoassay for one-step noncompetitive detection of corticosteroid 11-deoxycortisol.
Open-sandwich enzyme immunoassay for one-step noncompetitive detection of corticosteroid 11-deoxycortisol.
Analytical chemistry (2009-09-26)
Masaki Ihara, Tatsuya Suzuki, Norihiro Kobayashi, Junichi Goto, Hiroshi Ueda
ABSTRACT
A noncompetitive immunoassay has the potential for improved sensitivity and working range compared with corresponding competitive assays. However, monovalent antigens with less than 1000 in molecular weight are not susceptible to sandwich assays due to their small size. As a noncompetitive immunoassay that can be performed with a clone of an antibody, an open-sandwich immunoassay (OS-IA) based on the antigen-dependent stabilization of the antibody variable region (V(H) + V(L)) was applied to the quantification of 11-deoxycortisol (11-DC; M(r) 346.5), a corticosteroid serving as a diagnostic index for pituitary-adrenal function, as a model target hapten. By one step OS-IA detection of enzyme-labeled V(H) fragment bound to immobilized V(L) in the presence of sample in microplate wells, 11-DC was measured with a femtomolar detection limit and the working range was wider than that with corresponding competitive assay. In addition, the selectivity against analogues was found almost identical to that of conventional assays. The effect of the mutagenesis of a V(H) residue at the V(H)/V(L) interface to reduce background signal was also shown, implying the wider application of OS-IA in small molecule analyses.
Sigma-Aldrich
Reichstein′s substance S, ≥98%
Supelco
11-Deoxycortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®