MilliporeSigma
  • Interaction of antithyroid drugs with bovine serum albumin: electrophoretic and fluorimetric study.
Interaction of antithyroid drugs with bovine serum albumin: electrophoretic and fluorimetric study.
Journal of pharmaceutical sciences (2009-09-24)
Margarita S Chernov'yants, Andrey O Dolinkin, Anatoly V Chernyshev, Evgeniy V Khohlov, Elizaveta G Golovanova
ABSTRACT
The pre-equilibrium capillary zone electrophoretic (pre-eq CZE) method to determine association constants of active anionic forms of antithyroid drugs: 6-n-propyl-2-thiouracil (PTU), 6-methyl-2-thiouracil (MTU), 2-thiouracil (TU) with bovine serum albumin (BSA) under physiological pH 7.4 has been developed for the first time. Using the decrease of the selective electrochromatographic peak area of a drug anionic form due to binding with BSA the association constants K of the binary BSA complexes were calculated. It has been found that the binding constants (log K) of BSA with TU, MTU, and PTU are equal to 2.99, 1.85, and 2.11, respectively. The interaction of PTU, MTU, TU, 2-mercapto-1-methylimidazole (MMI), and ethyl-3-methyl-2-thionoimidazoline-1-carboxylate (Carb), which is considered to be a prodrug for MMI, with BSA has been investigated under physiological conditions by means of fluorescence spectroscopy. Fluorescence emission spectra of BSA in the presence of thioamides recorded at 295 nm excitation wavelength clearly show that the studied drugs act as quenchers, except MMI, which acts as quencher when being excited at 280 nm. The 295 nm light excites tryptophan residues, while the 280 nm light excites both tryptophan and tyrosine residues. The binding constants (log K) of BSA with PTU, MTU, TU, MMI, and Carb have been found to be 4.51, 4.30, 4.30, 2.64, and 4.32, respectively.
Product Description
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)