MilliporeSigma
  • Influence of curing protocol on selected properties of light-curing polymers: degree of conversion, volume contraction, elastic modulus, and glass transition temperature.
Influence of curing protocol on selected properties of light-curing polymers: degree of conversion, volume contraction, elastic modulus, and glass transition temperature.
Dental materials : official publication of the Academy of Dental Materials (2009-09-15)
Magali Dewaele, Erik Asmussen, Anne Peutzfeldt, E Christian Munksgaard, Ana R Benetti, Gauthier Finné, Gaëtane Leloup, Jacques Devaux
ABSTRACT
The purpose of this study was to investigate the effect of light-curing protocol on degree of conversion (DC), volume contraction (C), elastic modulus (E), and glass transition temperature (T(g)) as measured on a model polymer. It was a further aim to correlate the measured values with each other. Different light-curing protocols were used in order to investigate the influence of energy density (ED), power density (PD), and mode of cure on the properties. The modes of cure were continuous, pulse-delay, and stepped irradiation. DC was measured by Raman micro-spectroscopy. C was determined by pycnometry and a density column. E was measured by a dynamic mechanical analyzer (DMA), and T(g) was measured by differential scanning calorimetry (DSC). Data were submitted to two- and three-way ANOVA, and linear regression analyses. ED, PD, and mode of cure influenced DC, C, E, and T(g) of the polymer. A significant positive correlation was found between ED and DC (r=0.58), ED and E (r=0.51), and ED and T(g) (r=0.44). Taken together, ED and PD were significantly related to DC and E. The regression coefficient was positive for ED and negative for PD. Significant positive correlations were detected between DC and C (r=0.54), DC and E (r=0.61), and DC and T(g) (r=0.53). Comparisons between continuous and pulse-delay modes of cure showed significant influence of mode of cure: pulse-delay curing resulted in decreased DC, decreased C, and decreased T(g). Influence of mode of cure, when comparing continuous and step modes of cure, was more ambiguous. A complex relationship exists between curing protocol, microstructure of the resin and the investigated properties. The overall performance of a composite is thus indirectly affected by the curing protocol adopted, and the desired reduction of C may be in fact a consequence of the decrease in DC.
Sigma-Aldrich
Ethyl 4-(dimethylamino)benzoate, ≥99%
Supelco
Ethyl 4-(dimethylamino)benzoate, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland