MilliporeSigma
  • On the origins of kinetic resolution of cyclohexane-1,2-diols through stereoselective acylation by chiral tetrapeptides.
On the origins of kinetic resolution of cyclohexane-1,2-diols through stereoselective acylation by chiral tetrapeptides.
Organic letters (2009-09-02)
C B Shinisha, Raghavan B Sunoj
ABSTRACT
The relative energies of cyclohexane-1,2-diols and chiral tetrapeptide (2 (Boc) or 3 (Moc)) complexes calculated using DFT indicate a thermodynamic preference for chiral recognition toward (1R,2R)(e,e)-alpha isomer. The barrier for stereoselective acyl transfer is identified as lower for trans-(1R,2R)-cyclohexane-1,2-diol, leading to the kinetic resolution (KR) of trans-(1S,2S)-cyclohexane-1,2-diol. The prediction is in concert with the reported experiments for trans-diols, while that for hitherto unknown cis-diol demands experimental verification. It is proposed that desymmetrization would enable the resolution of cis-(1R,2S)-2-hydroxycyclohexyl acetate.
Sigma-Aldrich
cis-1,2-Cyclohexanediol, 99%
Sigma-Aldrich
trans-1,2-Cyclohexanediol, 98%