MilliporeSigma
  • Organocatalytic asymmetric Friedel-Crafts alkylation/cyclization cascade reaction of 1-naphthols and alpha,beta-unsaturated aldehydes: an enantioselective synthesis of chromanes and dihydrobenzopyranes.
Organocatalytic asymmetric Friedel-Crafts alkylation/cyclization cascade reaction of 1-naphthols and alpha,beta-unsaturated aldehydes: an enantioselective synthesis of chromanes and dihydrobenzopyranes.
The Journal of organic chemistry (2009-08-12)
Liang Hong, Lei Wang, Wangsheng Sun, Kwokyin Wong, Rui Wang
ABSTRACT
An enantioselective Friedel-Crafts alkylation/cyclization cascade reaction of 1-naphthols and alpha,beta-unsaturated aldehydes promoted by diphenylprolinol ether has been developed. The method affords one-pot access to chiral and synthetically useful chromanes and dihydrobenzopyranes in high yields and enantioselectivities from readily available compounds. In addition, the addition/cyclization products could be afterward transformed to various natural products and biologically active derivatives. On the basis of the experimental results and the observed absolute configurations of the products, a plausible mechanism has been proposed to explain the origin of the activation and the asymmetric induction.
Sigma-Aldrich
2,2,5,7,8-Pentamethyl-6-chromanol, 97%