Merck
  • Biological detection and analysis of toxicity of organophosphate- and azadirachtin-based insecticides in Lathyrus sativus L.
Biological detection and analysis of toxicity of organophosphate- and azadirachtin-based insecticides in Lathyrus sativus L.
Ecotoxicology (London, England) (2009-07-21)
Susmita Ganguly, Sima Bhattacharya, Sukumar Mandi, Jayanta Tarafdar
ABSTRACT
In this study, attention was paid to investigate the effect of organophosphate insecticides, profenofos 40% EC, methyl parathion (metacid) 50% EC, and neem-based product nimbecidine 0.03% EC (from Azadirachta indica) on somatic chromosomal behavior, level of leaf protein, and activity of antioxidant enzymes in Lathyrus sativus L., the leguminous herb. The experiments on somatic chromosomes of root tip cells of L. sativus L. revealed that most common type of abnormalities were anaphase bridge, chromosome fragment, breaks, giant interphase, etc. Also, the mitotic index reduced and abnormality index enhanced, which were directly proportional to the rise in concentration as well as time period of exposure of chemicals. The profenofos and metacid induced drastic changes in mitotic index when compared with nimbecidine. The electrophoretic studies of leaf protein of L. sativus L. showed alteration of some major and minor protein bands subjected to spraying of organophosphate insecticides and induced to synthesize additional high molecular mass protein compared to untreated control. Analysis of SOD, EST, and POD activity by non-denaturing polyacrylamide gel electrophoresis showed different patterns of the isoforms. Complete inhibition of EST was observed in profenofos-treated plants, while with metacid- and nimbecidine-treated plants EST was suppressed. Induction and/or increased activities of SOD and POD were generally enhanced. Our present study not only provides the important information for better understanding of the toxic and tolerance mechanisms, but as well can be used as a bio-indicator for contamination by pesticides, which could cause genetic instabilities of natural plant populations and in crop varieties.
Product Description
Supelco
Profenofos, PESTANAL®, analytical standard
Pricing and availability is not currently available.