MilliporeSigma
  • Cyclization of hydrazones of 2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene. Synthesis of Spiro naphthoxazine dimers.
Cyclization of hydrazones of 2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene. Synthesis of Spiro naphthoxazine dimers.
Molecules (Basel, Switzerland) (2009-06-26)
Abdullah Saad Al-Bogami, Abdullah Mohammed Al-Majid, Mohammed Ali Al-Saad, Ahmed Amine Mousa, Sara Abdullah Al-Mazroa, Hamad Zaid Alkhathlan
ABSTRACT
Cyclization of hydrazones derived from 2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene gave naphtho[1,2-e]-1,3-oxazines, naphtho[2,1-e]-1,3-oxazines or their spiro dimers depending on the molar ratio of triphosgene used for the cyclization.
Sigma-Aldrich
Bis(trichloromethyl) carbonate, purum, ≥99.0% (AT)
Sigma-Aldrich
Triphosgene, reagent grade, 98%