MilliporeSigma
  • Metabolism of acenaphthylene via 1,2-dihydroxynaphthalene and catechol by Stenotrophomonas sp. RMSK.
Metabolism of acenaphthylene via 1,2-dihydroxynaphthalene and catechol by Stenotrophomonas sp. RMSK.
Biodegradation (2009-06-23)
Anand S Nayak, Yaligara Veeranagouda, Kyoung Lee, T B Karegoudar
ABSTRACT
Stenotrophomonas sp. RMSK capable of degrading acenaphthylene as a sole source of carbon and energy was isolated from coal sample. Metabolites produced were analyzed and characterized by TLC, HPLC and mass spectrometry. Identification of naphthalene-1,8-dicarboxylic acid, 1-naphthoic acid, 1,2-dihydroxynaphthalene, salicylate and detection of key enzymes namely 1,2-dihydroxynaphthalene dioxygenase, salicylaldehyde dehydrogenase and catechol-1,2-dioxygenase in the cell free extract suggest that acenaphthylene metabolized via 1,2-dihydroxynaphthalene, salicylate and catechol. The terminal metabolite, catechol was then metabolized by catechol-1,2-dioxygenase to cis,cis-muconic acid, ultimately forming TCA cycle intermediates. Based on these studies, the proposed metabolic pathway in strain RMSK is, acenaphthylene --> naphthalene-1,8-dicarboxylic acid --> 1-naphthoic acid --> 1,2-dihydroxynaphthalene --> salicylic acid --> catechol --> cis,cis-muconic acid.
Product Description
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Acenaphthylene, 75%