MilliporeSigma
  • X-ray spectroscopic approaches to the investigation and characterization of photochemical processes.
X-ray spectroscopic approaches to the investigation and characterization of photochemical processes.
Journal of synchrotron radiation (2009-06-19)
Pierre Kennepohl, Erik C Wasinger, Serena DeBeer George
ABSTRACT
Despite a wealth of studies exemplifying the utility of the 2-5 keV X-ray range in speciation and electronic structure elucidation, the exploitation of this energy regime for the study of photochemical processes has not been forthcoming. Herein, a new endstation set-up for in situ photochemical soft X-ray spectroscopy in the 2-5 keV energy region at the Stanford Synchrotron Radiation Lightsource is described for continuous photolysis under anaerobic conditions at both cryogenic and ambient temperatures. Representative examples of this approach are used to demonstrate the potential information content in several fields of study, including organometallic chemistry, biochemistry and materials chemistry.
Sigma-Aldrich
N-Acetyl-L-methionine, ≥98.5% (T)
Sigma-Aldrich
N-Acetyl-D-methionine, ~99%