MilliporeSigma
  • Glycosidase inhibition by ring-modified castanospermine analogues: tackling enzyme selectivity by inhibitor tailoring.
Glycosidase inhibition by ring-modified castanospermine analogues: tackling enzyme selectivity by inhibitor tailoring.
Organic & biomolecular chemistry (2009-06-18)
Matilde Aguilar-Moncayo, Tracey M Gloster, Johan P Turkenburg, M Isabel García-Moreno, Carmen Ortiz Mellet, Gideon J Davies, José M García Fernández
ABSTRACT
Synthesis of a panel of iso(thio)urea-type ring-modified castanospermine analogues bearing a freely mutarotating pseudoanomeric hydroxyl group results in tight-binding beta-glucosidase inhibitors with unusual binding signatures; the presence of an N-octyl substituent imparts a remarkable anomeric selectivity, promoting strong binding of the appropriate beta-anomer by the beta-glucosidase.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%