MilliporeSigma
  • Light tolerance of R-phycoerythrin and a tandem conjugate observed by single molecule recrossing events.
Light tolerance of R-phycoerythrin and a tandem conjugate observed by single molecule recrossing events.
Applied spectroscopy (2009-06-18)
Sean M Burrows, Payal Patel, Dimitri Pappas
ABSTRACT
The recrossing of single molecules in a probe volume was used to investigate light harvesting and energy transfer between R-phycoerythrin (R-PE) and a tandem conjugate dye. The normalized recrossing ratio, Nr/Nt, was defined as the number of molecules that reenter the probe volume (Nr) to the total number of molecules detected (Nt). The energy transfer process in phycobiliproteins was studied as a function of excitation irradiation and irradiation time. This was achieved by investigating the average baseline-subtracted fluorescence intensity, normalized molecular recrossing ratio (Nr/Nt), and the number of molecules detected per second. The photon saturation irradiance of the R-PE and the tandem conjugate were compared with each other, showing that energy transfer to the tandem dye significantly improves photostability and light tolerance of the phycobiliprotein. The Nr/Nt ratio was used to study the photophysical properties of R-phycoerythrin and the tandem conjugate Streptavidin R-Phycoerythrin-AlexaFluor-647 (PE-647). Normalized molecular recrossings showed that energy transfer to a tandem conjugate could reduce the formation of triplet states in R-phycoerythrin and extend the light tolerance of certain phycobiliproteins.
Sigma-Aldrich
Streptavidin-R-Phycoerythrin, ≥1 mg/mL (active substance)
Sigma-Aldrich
8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate, ~98%