MilliporeSigma
Fractal aggregates evolution of methyl red in liquid crystal.
The European physical journal. E, Soft matter (2009-06-11)
F Ciuchi, L Sorriso-Valvo, A Mazzulla, J M Redondo
ABSTRACT
The spontaneous formation of dendritic aggregates is observed in a two-dimensional confined layered system consisting of a film composed of liquid crystal, dye and solvent cast above a polymer substrate. The observed aggregates are promoted by phase separation processes induced by dye diffusion and solvent evaporation. The growth properties of the aggregates are studied through the temporal evolution of their topological properties (surface, perimeter, fractal dimension). The fractal dimension of the completely formed structures, when they are coexistent with different types of structures, is consistent with theoretical and experimental values obtained for Diffusion-Limited Aggregates. Under different experimental conditions (temperature and local dye concentration) the structure forms without interactions with other kinds of structures, and its equilibrium fractal dimension is smaller. The fractal dimension is thus not a universal property of the observed structures, but rather depends on the experimental conditions.
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, indicator (pH 4.2-6.2)
Millipore
Methyl Red solution, suitable for microbiology