MilliporeSigma
Dy3+ and Nd3+ induced genetic mutation of bacillus alpha-amylase.
Journal of inorganic biochemistry (2009-06-06)
Dongyan Zhang, Jianhua Hu, Bater Siqin, Tong Zhang, Ying Chang
ABSTRACT
After treatment with DyCl(3) and NdCl(3), two strains of mutated bacilli showing increased alpha-amylase activity were isolated. The alpha-amylase genes were amplified, cloned, and sequenced. Sequencing revealed that there were either 11 or 14 bases altered in the two genes. These alterations took the form of base substitutions, and transversion was more common than transition. Based on these results, it was concluded that the rare earth compounds DyCl(3) and NdCl(3) were mutagens.
Sigma-Aldrich
Neodymium(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Pricing and availability is not currently available.