MilliporeSigma
  • Evaluation of pesticide toxicities with differing mechanisms using Caenorhabditis elegans.
Evaluation of pesticide toxicities with differing mechanisms using Caenorhabditis elegans.
Journal of toxicology and environmental health. Part A (2009-06-06)
Qin-Li Ruan, Jing-Juan Ju, Yun-Hui Li, Ran Liu, Yue-Pu Pu, Li-Hong Yin, Da-Yong Wang
ABSTRACT
The aim of this study was to (1) determine whether model organism Caenorhabditis elegans was sensitive to pesticides at the maximum concentration limits regulated by national agency standards, and (2) examine the multi-biological toxicities occurring as a result of exposure to pesticides. Five pesticides, namely, chlorpyrifos, imibacloprid, buprofezin, cyhalothrin, and glyphosate, with four different mechanisms of action were selected for the investigation. In accordance with national agency requirements, 4 exposed groups were used for each tested pesticide with the concentration scales ranging from 1.0 x 10(-3) to 1 mg/L. L4 larvae were exposed for 24 and 72 h, respectively. Endpoints of locomotion, propagation, and development were selected for the assay as parameters of toxicity. After exposure for 24 h, both the body bend frequency and head thrash frequency of nematodes exposed to chlorpyrifos, imibacloprid, and cyhalothrin decreased in a concentration-dependent manner, and there were significant differences between exposed groups at maximum concentration level (MCL) compared to control. The generation time of nematodes exposed to buprofezin 24 h significantly increased in a concentration-dependent manner in the highest exposed group. When exposed for 72 h, the body bend frequency and head thrash frequency of nematodes exposed to cyhalothrin markedly decreased at MCL. The generation time and brood size of nematodes exposed to buprofezin were reduced in a concentration-dependent manner. The behavior of nematodes was sensitive to pesticides with neurotoxic properties, while pesticides affecting insect growth modified the reproductive system. The effects of pesticides on nematodes exposed for 24 h appeared more sensitive than with exposure for 72 h. Caenorhabditis elegans may thus be used for assessing the adverse effects of pesticide residues in aquatic environment.
Product Description
Supelco
Buprofezin, PESTANAL®, analytical standard