MilliporeSigma
Substituent diversity-directed synthesis of indole derivatives.
Journal of combinatorial chemistry (2009-05-28)
Dong Mei Wang, Ming Na Sun, Gang Liu
ABSTRACT
This paper reports a versatile, good-yielding, solution-phase method that is a substituent diversity-directed synthesis of 1H-indoles (6-13, 17-20) and 1-hydroxyindoles (14, 15) starting from commercially available 1,5-difluoro-2,4-dinitrobenzene. The synthetic products possessed the maximum six diversity points.
Sigma-Aldrich
1,5-Difluoro-2,4-dinitrobenzene, 97%