MilliporeSigma
  • Identification of 2-aminobenzimidazoles as potent melanin-concentrating hormone 1-receptor (MCH1R) antagonists.
Identification of 2-aminobenzimidazoles as potent melanin-concentrating hormone 1-receptor (MCH1R) antagonists.
Bioorganic & medicinal chemistry letters (2009-05-22)
Minoru Moriya, Hiroyuki Kishino, Shunji Sakuraba, Toshihiro Sakamoto, Takuya Suga, Hidekazu Takahashi, Takao Suzuki, Masahiko Ito, Junko Ito, Ryuichi Moriya, Norihiro Takenaga, Hisashi Iwaasa, Akane Ishihara, Akio Kanatani, Takehiro Fukami
ABSTRACT
A series of 2-aminobenzimidazole-based MCH1R antagonists was identified by core replacement of the aminoquinoline lead 1. Subsequent modification of the 2- and 5-positions led to improvement in potency and intrinsic clearance. Compound 25 exhibited good plasma and brain exposure, and attenuated MCH induced food intake at 30mg/kg PO in rats.
Sigma-Aldrich
2-Aminobenzimidazole, 97%