MilliporeSigma
  • Attraction of New Zealand flower thrips, Thrips obscuratus, to cis-jasmone, a volatile identified from Japanese honeysuckle flowers.
Attraction of New Zealand flower thrips, Thrips obscuratus, to cis-jasmone, a volatile identified from Japanese honeysuckle flowers.
Journal of chemical ecology (2009-05-16)
A M El-Sayed, V J Mitchell, G F McLaren, L M Manning, B Bunn, D M Suckling
ABSTRACT
This work was undertaken to identify floral compound(s) produced by honeysuckle flowers, Lonicera japonica (Thunberg), that mediate the attraction of New Zealand flower thrips Thrips obscuratus (Crawford). Volatiles were collected during the day and night and analyzed by gas chromatography-mass spectrometry (GC-MS) to determine their emission over these two periods. Nine compounds were identified in the headspace; the main compound was linalool, and the other compounds were germacrene D, E,E-alpha-farnesene, nerolidol, cis-jasmone, cis-3-hexenyl acetate, hexyl acetate, cis-hexenyl tiglate, and indole. There was a quantitative difference between day and night volatiles, with cis-3-hexenyl acetate, hexyl acetate, cis-hexenyl tiglate, and cis-jasmone emitted in higher amounts during the day compared to the night. When the compounds were tested individually in field trapping experiments, only cis-jasmone attracted New Zealand flower thrips in a significant number. In another field trapping experiment, cis-jasmone caught similar numbers of New Zealand flower thrips compared to a floral blend formulated to mimic the ratios of the compounds emitted during the day, while catch with the night-emitted floral blend was not significantly different from the control. Subsequently, two field trapping experiments were conducted to determine the optimal attraction dose for cis-jasmone, a range of 1-100 mg loaded onto a red rubber stopper was tested, and the highest catches were in traps baited with 100 mg loading. A higher range of 100-1000 mg loaded into polyethylene vials was tested, and the highest catch was in traps baited with 500 mg. In another experiment aimed at comparing the attraction efficacy of cis-jasmone with the two other known thrips attractants (ethyl nicotinate and p-anisaldehyde), ethyl nicotinate showed the highest trap catch followed by cis-jasmone. A smaller number of Thrips tabaci (Lindeman) was attracted to traps baited with cis-jasmone. These results suggest that cis-jasmone might act as a kairomone that mediates the attraction of New Zealand flower thrips to the flowers of the Japanese honeysuckle.
Product Description
Supelco
Jasmone, analytical standard
Sigma-Aldrich
cis-Jasmone, stabilized