MilliporeSigma
Tinnitus as a prognostic factor of sudden deafness.
Acta oto-laryngologica (2009-05-14)
Noriko Hikita-Watanabe, Tadashi Kitahara, Arata Horii, Takayuki Kawashima, Katsumi Doi, Shin-Ichi Okumura
ABSTRACT
The 'tinnitus-rare' group had a poorer prognosis for hearing than the 'tinnitus-often' group in all sudden sensorineural hearing loss (SSNHL), although the 'shorter duration' group had better prognosis than the 'longer duration' when restricted to SSNHL accompanied by tinnitus. This indicates that tinnitus itself may not be a sign for poor hearing prognosis but might be an essential sound for the initiation of repair of a damaged auditory system. We examined the hearing improvement rate (HIR) and tinnitus at the onset of SSNHL to elucidate the prognostic value of tinnitus accompanying SSNHL. Fifty patients with SSNHL were treated with systemic administration of steroids. Hearing recovery was determined by comparing the hearing levels before and after treatment. Tinnitus was subjectively evaluated by the tinnitus scoring questionnaire. The score for the five-step evaluation of the subjective tinnitus feelings 'loudness', 'duration' and 'annoyance' was obtained at the onset. In terms of 'duration', when we divided all the cases into 'tinnitus-rare' group and 'tinnitus-often' group, HIR in the 'tinnitus-rare' group was significantly lower than that in 'tinnitus-often' group. When restricted to the 'tinnitus-often' group, HIR for 'shorter duration' was significantly higher than that for 'longer duration'.
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortisone 21-hemisuccinate sodium salt, powder, BioReagent, suitable for cell culture
Supelco
Hydrocortisone 21-hemisuccinate, analytical standard