MilliporeSigma
Selective antibacterial and apoptosis-modulating activities of mastic.
In vivo (Athens, Greece) (2009-05-06)
Hiroshi Sakagami, Kaori Kishino, Masaki Kobayashi, Ken Hashimoto, Satora Iida, Akiko Shimetani, Yuko Nakamura, Keiso Takahashi, Takeshi Ikarashi, Haruka Fukamachi, Kazue Satoh, Hideki Nakashima, Takahisa Shimizu, Ken Takeda, Shuji Watanabe, Wataru Nakamura
ABSTRACT
Mastic is a resinous exudate obtained from the stem and the main leaves of Pistacia lentiscus. We have reported the antiplaque effect of mastic-containing chewing gum on the oral cavity. We hypothesize that mastic may be a multifunctional food which has some beneficial pharmaceutical properties. The aim of this study was to assess the biological activity of solid and liquid types of mastic by cytotoxicity against fibroblasts, radical-scavenging activities and inhibitory effect on cell death of oral polymorphonuclear leukocytes (OPMNs). Mastic showed selective antibacterial action against Porphyromonas gingivalis and Prevotella melaninogenica, but no anti-HIV activity. Among a total of thirteen human cell types, promyelocytic leukemia HL-60 was the most sensitive to the cytotoxicity of mastic, followed by myeloblastic leukemia (ML-1, KG-1), erythroleukemia (K-562), oral squamous cell carcinoma (HSC-2, HSC-3, HSC-4), hepatocellular carcinoma (HepG2), glioblastoma (T98G, U87MG) and normal oral cells (gingival fibroblast, pulp cell, periodontal ligament fibroblast, most resistant). Mastic did not induce the differentiation of myelogenous leukemic cells into maturing cells with higher nitroblue tetrazolium-reducing activity, but induced apoptotic cell death, characterized by internucleosomal DNA fragmentation, caspase-3 activation and a decline in the intracellular concentration of putrescine. The cytotoxicity of mastic against leukemic cells did not diminish during its storage. On the other hand, mastic inhibited the spontaneous apoptosis of OPMNs. Mastic showed hydroxyl radical-scavenging activity. The selective antibacterial and apoptosis-modulating activity of mastic suggests its possible beneficial effects on oral health.
Sigma-Aldrich
Gum mastic