MilliporeSigma
  • Time-dependent density functional theory study on the electronic excited-state geometric structure, infrared spectra, and hydrogen bonding of a doubly hydrogen-bonded complex.
Time-dependent density functional theory study on the electronic excited-state geometric structure, infrared spectra, and hydrogen bonding of a doubly hydrogen-bonded complex.
Journal of computational chemistry (2009-04-29)
Yufang Liu, Junxia Ding, Ruiqiong Liu, Deheng Shi, Jinfeng Sun
ABSTRACT
The geometric structures and infrared (IR) spectra in the electronically excited state of a novel doubly hydrogen-bonded complex formed by fluorenone and alcohols, which has been observed by IR spectra in experimental study, are investigated by the time-dependent density functional theory (TDDFT) method. The geometric structures and IR spectra in both ground state and the S(1) state of this doubly hydrogen-bonded FN-2MeOH complex are calculated using the DFT and TDDFT methods, respectively. Two intermolecular hydrogen bonds are formed between FN and methanol molecules in the doubly hydrogen-bonded FN-2MeOH complex. Moreover, the formation of the second intermolecular hydrogen bond can make the first intermolecular hydrogen bond become slightly weak. Furthermore, it is confirmed that the spectral shoulder at around 1700 cm(-1) observed in the IR spectra should be assigned as the doubly hydrogen-bonded FN-2MeOH complex from our calculated results. The electronic excited-state hydrogen bonding dynamics is also studied by monitoring some vibraitonal modes related to the formation of hydrogen bonds in different electronic states. As a result, both the two intermolecular hydrogen bonds are significantly strengthened in the S(1) state of the doubly hydrogen-bonded FN-2MeOH complex. The hydrogen bond strengthening in the electronically excited state is similar to the previous study on the singly hydrogen-bonded FN-MeOH complex and play important role on the photophysics of fluorenone in solutions.
Sigma-Aldrich
9-Fluorenone, 98%