MilliporeSigma
  • Inhibition of subgenomic hepatitis C virus RNA replication by acridone derivatives: identification of an NS3 helicase inhibitor.
Inhibition of subgenomic hepatitis C virus RNA replication by acridone derivatives: identification of an NS3 helicase inhibitor.
Journal of medicinal chemistry (2009-04-25)
Giuseppe Manfroni, Jan Paeshuyse, Serena Massari, Samantha Zanoli, Barbara Gatto, Giovanni Maga, Oriana Tabarrini, Violetta Cecchetti, Arnaldo Fravolini, Johan Neyts
ABSTRACT
We report the synthesis and structure-activity relationship (SAR) of a large series of acridones and acridone-fragment derivatives designed on the basis of the selective antihepatitis C virus (HCV) activity shown by acridone 2, previously studied as a potential antibovine viral diarrhea virus (BVDV) compound. The evaluation of their ability to inhibit the HCV replication in Huh-5-2 cells led to the identification of new, selective inhibitors. This indicates that the acridone skeleton, when properly functionalized, is a suitable scaffold to obtain potential anti-HCV agents. Interestingly, during identification of possible cellular and viral targets, it was discovered that compound 23 exerts inhibitory activity on the HCV NS3 helicase, a very promising target for the development of anti-HCV drugs.
Sigma-Aldrich
9(10H)-Acridanone, 99%