MilliporeSigma
  • Generation of ortho-quinone methides by p-TsOH on silica and their hetero-Diels-Alder reactions with styrenes.
Generation of ortho-quinone methides by p-TsOH on silica and their hetero-Diels-Alder reactions with styrenes.
The Journal of organic chemistry (2009-04-24)
Paratchata Batsomboon, Wong Phakhodee, Somsak Ruchirawat, Poonsakdi Ploypradith
ABSTRACT
2-Arylchromans were readily prepared from the hetero-Diels-Alder reactions of styrenes with the ortho-quinone methides (o-QMs) which, in turn, were generated by treating the MOM-protected benzylacetate derivatives with p-TsOH immobilized on silica (PTS-Si) in toluene under mild conditions (0 degrees C to rt). The corresponding chromans were obtained in moderate to excellent yields (42-97%) and in moderate to excellent diastereoselectivity (up to >99:1).
Sigma-Aldrich
Sodium p-toluenesulfinate, 95%
Sigma-Aldrich
Sodium p-toluenesulfinate, purum, anhydrous, ≥96.0% (NT)