MilliporeSigma
Lanthanide(III)-doped magnetite nanoparticles.
Journal of the American Chemical Society (2009-04-17)
Channa R De Silva, Steve Smith, Inbo Shim, Jeffrey Pyun, Timothy Gutu, Jun Jiao, Zhiping Zheng
ABSTRACT
Nearly monodisperse lanthanide-doped magnetite nanoparticles were obtained by thermally decomposing a mixture of Fe(acac)(3) and Ln(acac)(3) (acac = acetylacetonate; Ln = Sm, Eu, Gd) in the presence of passivating surfactants. Magnetic studies revealed room-temperature ferromagnetic behaviors of these doped nanoparticles, distinctly different from those of the undoped parent magnetite or the doped nanoparticles prepared by a coprecipitation method.
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Manganese(II) acetylacetonate