MilliporeSigma
  • A validated HPLC-fluorescence method with a semi-micro column for routine determination of homocysteine, cysteine and cysteamine, and the relation between the thiol derivatives in normal human plasma.
A validated HPLC-fluorescence method with a semi-micro column for routine determination of homocysteine, cysteine and cysteamine, and the relation between the thiol derivatives in normal human plasma.
Biomedical chromatography : BMC (2009-04-09)
Saori Ichinose, Mayumi Nakamura, Mayu Maeda, Rie Ikeda, Mitsuhiro Wada, Mio Nakazato, Yoshihito Ohba, Noboru Takamura, Takahiro Maeda, Kiyoshi Aoyagi, Kenichiro Nakashima
ABSTRACT
A semi-micro column HPLC-fluorescence method for routine determination of thiol derivatives such as homocysteine (Hcy), cysteine (Cys) and cysteamine (CA) is described. The thiol derivatives labeled with ammonium-7-fluorobenzo-2-oxa-1,3-diazole-4-sulfonate (SBD-F) were isocratically separated within 12 min on a semi-micro ODS column (Daisopak-SP-120-5-ODS-BP) with a mixture of 25 mm acetate buffer (pH 2.00) and CH(3)CN as a mobile phase. The purity and similarity of SBD-thiols by a multi-wavelength fluorescence detector were more than 92.3 and 96.7%. The detection limits of Hcy, Cys and CA at a signal-to-noise ratio of 3 were 0.16, 0.47 and 0.03 microm, respectively. Furthermore validation parameters such as accuracy, precision and robustness of the proposed method showed satisfactory results. Almost 850 plasma sample injections (range 572-1076, n = 3) for a column could be performed without differences in retention time and peak heights of labels. As an application of the proposed method, the determination of thiol derivatives in normal human plasma (n = 103) was demonstrated. The correlation coefficients between Hcy vs Cys and Hcy vs CA were 0.38 and -0.35, respectively.
Product Description
Supelco
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, for HPLC derivatization, LiChropur, ≥98.5% (HPLC)
Sigma-Aldrich
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, ≥98%