MilliporeSigma
  • Energetics and kinetics of photosynthetic water oxidation studied by photothermal beam deflection (PBD) experiments.
Energetics and kinetics of photosynthetic water oxidation studied by photothermal beam deflection (PBD) experiments.
Photosynthesis research (2009-03-31)
André Klauss, Roland Krivanek, Holger Dau, Michael Haumann
ABSTRACT
Determination of thermodynamic parameters of water oxidation at the photosystem II (PSII) manganese complex is a major challenge. Photothermal beam deflection (PBD) spectroscopy determines enthalpy changes (ΔH) and apparent volume changes which are coupled with electron transfer in the S-state cycle (Krivanek R, Dau H, Haumann M (2008) Biophys J 94: 1890–1903). Recent PBD results on formation of the Q⁻(A)/Y(•+)(Z) radical pair suggest a value of ΔH similar to the free energy change, ΔG, of -540±40 meV previously determined by the analysis of recombination fluorescence, but presently the uncertainty range of ΔH values determined by PBD is still high (±250 meV). In the oxygen-evolving transition, S₃−−>S₀, the enthalpy change may be close to zero. A prominent non-thermal signal is associated with both Q⁻(A)/Y(•+)(Z) formation (<1 μs) and the S₃−−>S₀ transition (~1 ms). The observed (apparent) volume expansion (ΔV of about +40 ų per PSII unit) in the S₃−−>S₀ transition seems to revert, at least partially, the contractions on lower S-transitions and may also comprise contributions from O₂ and proton release. The observed volume changes show that the S₃−−>S₀ transition is accompanied by significant nuclear movements, which likely are of importance with respect to energetics and mechanism of photosynthetic water oxidation. Detailed PBD studies on all S-transitions will contribute to the progress in PSII research by providing insights not accessible by other spectroscopic methods.
Sigma-Aldrich
Bromocresol Purple, Technical grade
Sigma-Aldrich
Bromocresol Purple, BioReagent, suitable for indicator, Dye content 90 %