MilliporeSigma
  • Grandine A, a new proaporphine alkaloid from the bark of Phoebe grandis.
Grandine A, a new proaporphine alkaloid from the bark of Phoebe grandis.
Molecules (Basel, Switzerland) (2009-03-28)
Mat Ropi Mukhtar, Ahmad Nazif Aziz, Noel F Thomas, A Hamid A Hadi, Marc Litaudon, Khalijah Awang
ABSTRACT
The stem bark of Phoebe grandis afforded one new oxoproaporphine; (-)-grandine A (1), along with six known isoquinoline alkaloids: (-)-8,9-dihydrolinearisine (2), boldine, norboldine, lauformine, scortechiniine A and scortechiniine B. In addition to that of the new compound, complete 1H- and 13C-NMR data of the tetrahydroproaporphine (-)-8,9-dihydrolinearisine (2) is also reported. The alkaloids' structures were elucidated primarily by means of high field 1D- and 2D-NMR and HRMS spectral data.
Product Description
Supelco
Boldine, analytical standard