MilliporeSigma
  • Oxoammonium salt/NaClO2: an expedient, catalytic system for one-pot oxidation of primary alcohols to carboxylic acids with broad substrate applicability.
Oxoammonium salt/NaClO2: an expedient, catalytic system for one-pot oxidation of primary alcohols to carboxylic acids with broad substrate applicability.
Chemical communications (Cambridge, England) (2009-03-19)
Masatoshi Shibuya, Takahisa Sato, Masaki Tomizawa, Yoshiharu Iwabuchi
ABSTRACT
A facile, green, one-pot oxidation of primary alcohols to carboxylic acids with broad substrate applicability has been developed by employing an expedient catalytic system consisting of 1-Me-AZADO+X-/NaClO2.
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%