MilliporeSigma
  • Bicarbonate-dependent and bicarbonate-independent mechanisms contribute to nondiffusive uptake of acetate in the ruminal epithelium of sheep.
Bicarbonate-dependent and bicarbonate-independent mechanisms contribute to nondiffusive uptake of acetate in the ruminal epithelium of sheep.
American journal of physiology. Gastrointestinal and liver physiology (2009-03-07)
Jörg R Aschenbach, Sabine Bilk, Gebrehiwot Tadesse, Friederike Stumpff, Gotthold Gäbel
ABSTRACT
The present study investigated the significance of apical transport proteins for ruminal acetate absorption and their interaction with different anions. In anion competition experiments in the washed reticulorumen, chloride disappearance rate (initial concentration, 28 mM) was inhibited by the presence of a short-chain fatty acid mixture (15 or 30 mM of each acetate, propionate, and butyrate). Disappearance rates of acetate and propionate, but not butyrate (initial concentration, 25 mM each) were diminished by 40 or 80 mM chloride. In isolated ovine ruminal epithelia mounted in Ussing chambers, an increase in chloride concentration from 4.5 to 90 mM led to a decrease of apical acetate uptake at a concentration of 0.5 mM. Mucosal nitrate inhibited acetate uptake most potently whereas sulfate had no effect. Decreasing mucosal pH from 7.4 to 6.1 approximately doubled uptake of acetate both at 0.5 and 10 mM, but this doubling was almost abolished when HCO(3)(-) was absent. The stimulated uptake at mucosal pH 6.1 consisted of a bicarbonate-dependent, nitrate-inhibitable part (K(m) = 54 mM) and a bicarbonate-independent component (K(m) = 12 mM) that was also sensitive to nitrate inhibition. Maximal uptake was three times larger for bicarbonate-dependent vs. bicarbonate-independent uptake. Mucosal addition of 200 microM DIDS, 400 microM p-chloromercuribenzene sulfonic acid, 800 microM p-hydroxymercuribenzoic acid, or 100 microM phloretin had no effects on acetate uptake although the latter two inhibited l-lactate uptake. Our data conclusively show a dominant involvement of proteins in apical acetate uptake. Previously described pH effects on acetate absorption originate mainly from modulation of acetate/bicarbonate exchange. Additionally, there is bicarbonate-independent uptake of acetate anions that is protein coupled but not via monocarboxylate cotransporter.
Sigma-Aldrich
4-(Hydroxymercuri)benzoic acid sodium salt, ≥95.0% (Hg)