MilliporeSigma
  • Flow injection chemiluminescence sensor using molecularly imprinted polymers as recognition element for determination of maleic hydrazide.
Flow injection chemiluminescence sensor using molecularly imprinted polymers as recognition element for determination of maleic hydrazide.
Biosensors & bioelectronics (2009-03-06)
Yanjun Fang, Shoulei Yan, Baoan Ning, Nan Liu, Zhixian Gao, Fuhuan Chao
ABSTRACT
A novel flow injection chemiluminescence (CL) sensor for the determination of maleic hydrazide (MH) using molecularly imprinted polymer (MIP) as recognition element is reported. The MH-MIP was synthesized by thermal initiated polymerization in methanol, using methacrylic acid (MAA) as functional monomer and ethylene glycol dimethacrylate (EGDMA) as cross-linker in the maleic hydrazide template molecule. Molecular modeling was employed to simulate the possible recognition process of the MIP, and to achieve high selectivity. Then the synthesized MH-MIP was employed as recognition element by packing into flow cell to establish a novel flow injection CL sensor. The CL intensity responded linearly to the concentration of MH in the range 3.5x10(-4)-5.0x10(-2) mg/mL with a detection limit of 6.0x10(-5) mg/mL (3sigma), which is lower than that of conventional methods. The relative standard deviation (RSD) for the determination of 1.0 microg/mL of MH was 2.7% (n=11). The sensor is reusable and has a great improvement in sensitivity and selectivity for CL analysis. As a result, the new MIP-CL sensor had been successfully applied to the determination of MH in vegetable samples.
Sigma-Aldrich
Maleic hydrazide, ReagentPlus®, 99%